Anti-ACTH 18-39 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0442R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ACTH 18-39
- Antigen symbol:ACTH 18-39
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:18-39/39
- Form:Liquid
- Gene ID:5443
- Antigen synonyms:MSH|NPP|POC|CLIP|LPH|Corticotropin-lipotropin|POMC|ACTH|Pro-opiomelanocortin
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:2.5/4.5 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ACTH
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
ACTH stimulates the adrenal glands to release cortisol. MSH (melanocyte-stimulating hormone) increases the pigmentation of skin by increasing melanin production in melanocytes. Beta-endorphin and Met-enkephalin are endogenous opiates.
Type: Primary
Antigen: ACTH 18-39
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 18-39/39
Host: Rabbit
Isotype: IgG
Reactivity: Rat