Anti-Myelin Protein Zero Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-0337R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Myelin Protein Zero
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Dog,Rat,Mouse
- Western blot:Yes
- Epitope:175-248/248
- Form:Liquid
- Gene ID:4359
- Antigen synonyms:CHM|Myelin protein P|P|CMT1B|Myelin peripheral protein|CMT4E|Myelin protein zero|MPZ|CMTDID|HMSNIB|CMT2I|MPP|CMTDI3|CMT2J|DSS|CMT1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:24 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Myelin Protein Zero
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Creation of an extracellular membrane face which guides the wrapping process and ultimately compacts adjacent lamellae.
Type: Primary
Antigen: Myelin Protein Zero
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 175-248/248
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Dog