Order Entry
Export
ContactUsLinkComponent
 
Anti-C Peptide Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-0274R-A680
 :  Bioss
Anti-C Peptide Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-0274R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    C Peptide
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    57-87/110
  • Form:
    Liquid
  • Gene ID:
    3630
  • Antigen synonyms:
    Hyperproinsulinemia|INS|C-Peptide|proinsulin precursor|Propeptide|INS_HUMAN.|Proinsulin|Insulin Precursor|IRDN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    3 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide of human C Peptide
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

C Peptide is part of the molecule of Proinsulin, that consists of three parts: C Peptide and two long strands of amino acids (called the alpha and beta chains) that later become linked together to form the insulin molecule. From every molecule of proinsulin, one molecule of insulin plus one molecule of C Peptide are produced. C peptide is released into the blood stream in equal amounts to insulin. A test of C peptide levels will show how much insulin the body is making. Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver.

Type: Primary
Antigen: C Peptide
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 57-87/110
Host: Rabbit
Isotype: IgG
Reactivity: