Anti-PACAP-27/38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-0190R-A750
: Bioss
- Antibody type:Primary
- Antigen name:PACAP-27/38
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:132-169/176
- Form:Liquid
- Gene ID:116
- Antigen synonyms:PACAP-38|PACAP-27|PACAP|Pituitary adenylate cyclase-activating polypeptide|ADCYAP1|PACAP27|PACAP38.
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:19 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PACAP-27/38
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Binding to its receptor activates G proteins and stimulates adenylate cyclase in pituitary cells. Promotes neuron projection development through the RAPGEF2/Rap1/B-Raf/ERK pathway.
Type: Primary
Antigen: PACAP-27/38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 132-169/176
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat