Order Entry
Export
ContactUsLinkComponent
Anti-PACAP-27/38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-PACAP-27/38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0190R-A680
Supplier:  Bioss
Anti-PACAP-27/38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0190R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PACAP-27/38
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    132-169/176
  • Form:
    Liquid
  • Gene ID:
    116
  • Antigen synonyms:
    PACAP-38|PACAP-27|PACAP|Pituitary adenylate cyclase-activating polypeptide|ADCYAP1|PACAP27|PACAP38.
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    19 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human PACAP-27/38
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Binding to its receptor activates G proteins and stimulates adenylate cyclase in pituitary cells. Promotes neuron projection development through the RAPGEF2/Rap1/B-Raf/ERK pathway.

Type: Primary
Antigen: PACAP-27/38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 132-169/176
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat