Anti-beta Amyloid 1-42 Mouse Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0076M-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:beta Amyloid 1-42
- Antigen symbol:beta Amyloid 1-42
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:35-42/42
- Form:Liquid
- Gene ID:320
- Antigen synonyms:X11ALPHA|MINT1|Neuronal Munc18-1-interacting protein 1|Amyloid beta A4 precursor protein-binding family A member 1|Neuron-specific X11 protein|APBA1|Adapter protein X11alpha|LIN1|X11|X11A|Mint-1|D9S411E
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:4,4 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human beta-Amyloid 1-42
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Putative function in synaptic vesicle exocytosis by binding to Munc18-1, an essential component of the synaptic vesicle exocytotic machinery. May modulate processing of the beta-amyloid precursor protein (APP) and hence formation of beta-APP.
Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 35-42/42
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse