Anti-ChAT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-0042R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Choline O-acetyltransferase
- Antigen symbol:ChAT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Epitope:101-200/748
- Form:Liquid
- Gene ID:1103
- Antigen synonyms:Choline O-acetyltransferase|CMS1A2|CHOACTASE|Choline acetylase|ChAT|CMS1A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:82 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ChAT
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
catalyses the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
Type: Primary
Antigen: Choline O-acetyltransferase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/748
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog