Order Entry
Export
ContactUsLinkComponent
Anti-OVCA1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-OVCA1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5714R-A750
Supplier:  Bioss
Anti-OVCA1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5714R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    OVCA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    251-350/443
  • Form:
    Liquid
  • Gene ID:
    1801
  • Antigen synonyms:
    DPH1_HUMAN|DPH1|Candidate tumor suppressor in ovarian cancer 1|DPH2L1|Diphthamide biosynthesis protein 2 like|Ovarian cancer associated gene 1 protein|Diptheria toxin resistance protein required for diphthamide biosynthesis like 1|DPH like 1|Diphthamide biosynthesis protein 1|Diphthamide biosynthesis protein 2 homolog like 1|DPH1 homolog S. cerevisiae|DPH2 like 1|DPH1 homolog|Diptheria toxin resistance protein required for diphthamide biosynthesis Saccharomyces like 1 |DPH like 1 S. cerevisiae|Ovarian tumor suppressor candidate 1|HsDph1|Ovarian cancer associated gene 1|DPH2L
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    49 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human OVCA1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

OVCA1 is required for the first step in the synthesis of diphthamide, a unique posttranslationally modified histidine found only in translation elongation factor-2 (EEF2; MIM 130610). This modification is conserved from archaebacteria to humans and serves as the target for ADP-ribosylation and inactivation of EEF2 by diphtheria toxin (DT) and Pseudomonas exotoxin A. OVCA1 is one of several enzymes involved in synthesis of diphthamide in EEF2.

Type: Primary
Antigen: OVCA1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/443
Host: Rabbit
Isotype: IgG
Reactivity: Human