Order Entry
Export
ContactUsLinkComponent
Anti-MEK1/2 Ser222 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-MEK1/2 Ser222 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-3722R-A680
 :  Bioss
Anti-MEK1/2 Ser222 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-3722R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    MEK1/2 Ser222
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    5604
  • Antigen synonyms:
    MKK 1|Dual specicity mitogen activated protein kinase kinase 1|Mitogen activated protein kinase kinase 1|MAPKK 1|Protein kinase mitogen activated kinase 1|Protein kinase mitogen activated, kinase 1|MAPKK1|MP2K1_HUMAN|MAP kinase/Erk kinase 1|MEK 1|Protein kinase mitogen activated kinase 1 MAP kinase kinase 1|PRKMK 1|ERK activator kinase 1|PRKMK1|MEK1Ser222|Map2K1|MAP kinase kinase 1|MAPK/ERK kinase 1|MEKK1|MKK1|MAP/Erk kinase 1|p-MEK-1/2Ser222
  • Modification:
    Phosphorylation (Ser222)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    42 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human MEK1/2 around the phosphorylation site of Ser222
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

The protein encoded by this gene is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development.

Type: Primary
Antigen: MEK1/2 Ser222
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat