Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-2980R-A680
: Bioss
- Antibody type:Primary
- Antigen name:FUS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:421-526/526
- Form:Liquid
- Gene ID:2521
- Antigen synonyms:HNRNPP2|TLS|Oncogene TLS|FUS1|ALS6|Translocated in liposarcoma protein|ETM4|75 kDa DNA-pairing protein|FUS|POMP75|RNA-binding protein FUS|Oncogene FUS
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TLS
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 421-526/526
Host: Rabbit
Isotype: IgG
Reactivity: Rat