Order Entry
Export
ContactUsLinkComponent
Anti-HBeAg Mouse Monoclonal Antibody [clone: 2E9] (HRP (Horseradish Peroxidase))
Anti-HBeAg Mouse Monoclonal Antibody [clone: 2E9] (HRP (Horseradish Peroxidase))
Catalog # BOSSBSM-2022M-HRP
Supplier:  Bioss
Anti-HBeAg Mouse Monoclonal Antibody [clone: 2E9] (HRP (Horseradish Peroxidase))
Catalog # BOSSBSM-2022M-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HBeAg
  • Clonality:
    Monoclonal
  • Clone:
    2E9
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    External core antigen|HBeAg|Core protein|Hepatitis B|HBe antigen|Precore/core|Precore protein|Core antigen|Pre C,C|Precore/core protei|HBVgp4|HBEAG_HBVD3|Precore/core ORF|p25
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    HBeAg full length protein
  • Purification:
    Purified by Protein G
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Hepatitis B e-antigen (HBeAg) is a viral protein associated with HBV infections. Unlike the surface antigen, the e-antigen is found in the blood only when there are viruses also present. When the virus goes into hiding,? the e-antigen will no longer be present in the blood. HBeAg is often used as a marker of ability to spread the virus to other people (infectivity). Measurement of e-antigen may also be used to monitor the effectiveness of HBV treatment; successful treatment will usually eliminate HBeAg from the blood and lead to development of against e-antigen (anti-HBe). There are some types (strains) of HBV that do not make e-antigen; these are especially common in the Middle East and Asia. In areas where these strains of HBV are common, testing for HBeAg is not very useful.

Type: Primary
Antigen: HBeAg
Clonality: Monoclonal
Clone: 2E9
Conjugation: HRP
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Virus