Order Entry
Export
ContactUsLinkComponent
Anti-Proteasome 19S S5A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Proteasome 19S S5A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8322R-A750
Supplier:  Bioss
Anti-Proteasome 19S S5A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-8322R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome 19S S5A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    101-200/377
  • Form:
    Liquid
  • Gene ID:
    5710
  • Antigen synonyms:
    AF1|AF|Antisecretory factor 1|S5a/antisecretory factor protein|Proteasome Prosome macropain 26S subunit non ATPase 4|ASF|Rpn 10|S5A|pUBR5|PSMD 4|MCB 1|Rpn10|26S proteasome non ATPase regulatory subunit 4|pUB R5|26S protease subunit S5a|26S proteasome regulatory subunit S5A|Prosome macropain|Multiubiquitin chain-binding protein|Multiubiquitin chain binding protein|Proteasome 26S subunit non ATPase 4|PSMD4|MCB1|26S proteasome non-ATPase regulatory subunit 4|PSMD4_HUMAN|Proteasome 26S non ATPase subunit 4|DS5a|26S proteasome regulatory subunit rpn10|AF 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    41 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Proteasome 19S S5A
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Binds and presumably selects ubiquitin-conjugates for destruction. Displays selectivity for longer polyubiquitin chains. Modulates intestinal fluid secretion.

Type: Primary
Antigen: Proteasome 19S S5A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/377
Host: Rabbit
Isotype: IgG
Reactivity: Mouse