Anti-RTF1 Rabbit Polyclonal Antibody
NOVUNB100-61052
- Antibody type:Primary
- Antigen name:RTF1 homolog, Paf1/RNA polymerase II complex component
- Antigen symbol:RTF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:23168
- Antigen synonyms:KIAA0252GTL7|RNA polymerase-associated protein RTF1 homolog|homolog (S. cerevisiae)|ortholog of mouse gene trap locus 7|Rtf1|Paf1/RNA polymerase II complex component
- Storage buffer:TBS and 0,1% BSA
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:0,2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to the C-terminus of human Restores TBP (TATA box-binding protein) Function-1 using the numbering given in entry EAW92492.1 (GeneID 23168).
- Purification:Immunogen affinity purified
- Pk:100 µl
The RTF1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RTF1. This antibody reacts with human, mouse. The RTF1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 to 1:500
Type: Primary
Antigen: RTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse