jo_SPEEDY S1P ESD Safety Sandals
Supplier: ELTEN
Certificates
About this item
Safety sandal, jo_SPEEDY, in black/blue finish.
- Mesh/microfibre material
- Fabric lining for natural ventilation
- Padded tongue
- Full-length inlay sole ESD
- Non-metallic, penetration-resistant midsole
- Rubber/EVA sole jo_DYNAMIC
- Composite toe cap
For dry work areas, applications including industry, storage, transport, assembly etc. For areas where there is a risk of penetration from pointed and sharp objects (S1P) or areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certifications: EN ISO 20345 S1P HRO/SRC, form A.
Specifications
- Catalog No:
- ELTN11171/39
- ELTN11171/42
- ELTN11171/43
- ELTN11171/40
- ELTN11171/41
- ELTN11171/46
- ELTN11171/36
- ELTN11171/47
- ELTN11171/44
- ELTN11171/45
- ELTN11171/37
- ELTN11171/48
- ELTN11171/38
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001