NELSON XXG GTX S3 HI CI ESD Safety Shoes, Black, Low
Supplier: ELTEN
Certificates
About this item
Safety shoe, NELSON XXG GTX, lace-up in black finish.
- Hydrophobized nubuck leather/hydrophobized textile material
- GORE-TEX performance comfort footwear
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO black
- Non-metallic, penetration-resistant midsole
- Rubber/PU sole WELLMAXX GRIP
- Composite toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certifications: EN ISO 20345 S3 HI/CI/HRO/SRC, form A.
Specifications
- Catalog No:
- ELTN728691/40
- ELTN728691/43
- ELTN728691/44
- ELTN728691/41
- ELTN728691/42
- ELTN728691/47
- ELTN728691/48
- ELTN728691/45
- ELTN728691/46
- ELTN728691/39
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001