Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,4-dibromo-6-fluorobenzoate
Methyl 2,4-dibromo-6-fluorobenzoate ≥97%
Methyl 2,4-dibromo-6-fluorobenzoate ≥97%
:
Apollo Scientific
Methyl 2,4-dibromo-6-fluorobenzoate ≥97%
:
Apollo Scientific
:
773134-10-4
:
:
C₈H₅Br₂FO₂
:
MFCD06204165
:
311.9305 g/mol