Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-4-fluoro-2-nitroaniline
3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%
3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%
3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dichloro-4-fluoro-2-nitroaniline ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1698952-59-8
:
Formula:
C₆H₃Cl₂FN₂O₂
MDL Number:
MFCD30162392
:
225.00462 g/mol