Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Hydroxy-4H-isochromene-1,3-dione
8-Hydroxy-4H-isochromene-1,3-dione ≥95%
8-Hydroxy-4H-isochromene-1,3-dione ≥95%
8-Hydroxy-4H-isochromene-1,3-dione ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
8-Hydroxy-4H-isochromene-1,3-dione ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
19723-36-5
:
Formula:
C₉H₆O₄
MDL Number:
MFCD24716986
:
178.14154 g/mol