Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Methylthio)pyrimidine-4,6-diol
2-(Methylthio)pyrimidine-4,6-diol 98+%
2-(Methylthio)pyrimidine-4,6-diol 98+%
2-(Methylthio)pyrimidine-4,6-diol 98+%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(Methylthio)pyrimidine-4,6-diol 98+%
Supplier:
Apollo Scientific
About this item
CAS Number:
29639-68-7
:
Formula:
C₅H₆N₂O₂S
MDL Number:
MFCD00006087
:
158.18 g/mol