ContactUsLinkComponent
Digital transmitted light microscopes, Mateo TL
Digital transmitted light microscopes, Mateo TL
Supplier:  LEICA MICROSYSTEMS
Certificates
undefined
Digital transmitted light microscopes, Mateo TL
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Catalog No:
  • 630-3514
  • 630-3515
  • 630-3488
  • 630-3513
  • Nosepiece:
  • Quadruple
  • Quadruple
  • Stage:
  • Fixed, 262L×212W mm
  • Fixed, 262L×212W mm
  • Condenser:
  • S40/0,45, 3-position slider for S40 condenser (BF, PH0, PH1)
  • S40/0,45, 3-position slider for S40 condenser (BF, PH0, PH1)
  • Camera:
  • 6 MP colour
  • 6 MP colour
  • Display:
  • 396,24 mm (15,6"), 1920×1080 monitor
  • 396,24 mm (15,6"), 1920×1080 monitor
  • Output:
  • 1× USB 3.0 port and 2× USB 2.0 ports
  • 1× USB 3.0 port and 2× USB 2.0 ports
Specifications