2,3,3'-Trichlorobiphenyl (PCB No. 20), Sigma-Aldrich®
Catalog # SIALBCR290-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
ADR:9,II
:257.55 g/mol
UN:3432
CAS Number:38444-84-7
Formula:C₁₂H₇Cl₃