Betulin ≥98.0% (by HPLC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:473-98-3
:
Formula:C₃₀H₅₀O₂
MDL Number:MFCD00016802
:442.73 g/mol
Melting Pt:256…257°C
Storage Temperature:Refrigerator
EINECS:207-475-5
Specification Test Results
- Appearance:White to light yelow to light beige powder
- Infrared spectrum:Conforms
- HPLC:>=98.0%
- Loss on drying:=<5.0%