1,2:7,8-Diepoxyoctane 98%
Catalog #
Jump to:
Product Details & Documents
Storage condition: Inert atmosphere, Room Temperature
Synonyms: 1,4-Di(oxiran-2-yl)butane
- CAS Number:2426-07-5
- Formula:C₈H₁₄O₂
- MDL Number:MFCD00005155
- Molecular Weight:142.20 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Docking Protein 2
- Cat. No.:USBID8080-02K
- Reactivity:Human
- Conjugation:Unconjugated
- Antigen symbol:DOK2
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.



