Potassium sulphate
Catalog # SIALP0772-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle, wide mouth
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:174.26 g/mol
Merck Index:13,07759
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-915-5
CAS Number:7778-80-5
Formula:K₂SO₄
MDL Number:MFCD00011388