

3-Mercapto-1,2-propanediol NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:96-27-5
:
Formula:C₃H₈O₂S
MDL Number:MFCD00004879
:108.16 g/mol
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- ASSAY (ANHYDROUS BASIS):97.0 - 101.0 %
- SPECIFIC GRAVITY:1.241 - 1.250
- REFRACTIVE INDEX:1.521 - 1.526
- pH OF A 1 IN 10 SOLUTION:3.5 - 7.0
- RESIDUE ON IGNITION:Max. 0.1 %
- SELENIUM:Max. 30 µg/g
- ELEMENTAL IMPURITIES:AS REPORTED
- WATER DETERMINATION:Max. 5.0 %
- IDENTIFICATION:TO PASS TEST
- RESIDUAL SOLVENTS:AS REPORTED
Specification Test Results
Related Information
M4080, Monothioglycerol, NF Sceintific Documnetation M4080, Monothioglycerol, NF Technical Details