Order Entry
Export
ContactUsLinkComponent
n-Vinylpyrrolidone/Styrene copolymer 38%, aqueous emulsion
n-Vinylpyrrolidone/Styrene copolymer 38%, aqueous emulsion
Catalog # SPCMV1019-250ML
Supplier:  Spectrum Chemical
CAS Number:  
n-Vinylpyrrolidone/Styrene copolymer 38%, aqueous emulsion
Catalog # SPCMV1019-250ML
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • pH:
    2.0 - 5.0
  • Viscosity (25°C, BrooKField):
    200.0 - 800.0 cPs
  • Solid Content:
    38.0 - 42.0 %

Specification Test Results

About this item

 :
Density:1.04
Storage Temperature:Ambient
MDL Number:MFCD00145020