Order Entry
Export
Orders LinkContactUsLinkComponent
 

Perchloric acid ≥70%

Oxidizing
Corrosive
Supplier:   Merck
Perchloric acid ≥70%
Supplier:  Merck
Certificates
Perchloric acid ≥70%

2 Options Available Below

Product Details & Documents
  • CAS Number:7601-90-3
  • Formula:HClO₄
  • MDL Number:MFCD00011325
  • Molecular Weight:100.46 g/mol
  • Boiling Pt:160…181 °C (1013 hPa)
  • Melting Pt:–112.2 °C
  • Density:1.53…1.768 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-512-4
  • UN:1873
  • ADR:5.1,I

 

Specifications for Product Family

Specification Test Results

  • Assay:
    70%
Product Family Options

Product Information