
Pyrrolo[2,1-f][1,2,4]triazin-4-amine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:159326-68-8
:
Formula:C₆H₆N₄
MDL Number:MFCD08234647
:134.14 g/mol
:GHS07
Specification Test Results
- HPLC:≥94 %
- Appearance (Color):Almost white to grey
- Appearance (Form):Powder
- Infrared spectrum:Conforms