Order Entry
Export
ContactUsLinkComponent
Molybdic acid ≥85% for synthesis
Molybdic acid ≥85% for synthesis
Catalog # 91302.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Molybdic acid ≥85% for synthesis
Catalog # 91302.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min 85%

Specification Test Results

About this item

 :
Merck Index:13,06262
Boiling Pt:∼300 °C (1013 hPa)
Melting Pt:300 °C
Density:3.1 g/cm³ (20 °C)
 :161.95 g/mol
Storage Temperature:Refrigerator
EINECS:231-970-5
CAS Number:7782-91-4
Formula:H₂MoO₄
MDL Number:MFCD00036287