Order Entry
Export
ContactUsLinkComponent
Sulpho-LC-SDA (Sulpho-NHS-LC-Diazirine) (Sulphosuccinimidyl 6-(4,4′-azipentanamido)hexanoate) ≥90%, Sigma-Aldrich®
Irritant
Sulpho-LC-SDA (Sulpho-NHS-LC-Diazirine) (Sulphosuccinimidyl 6-(4,4′-azipentanamido)hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803359-50MG
Supplier:  Merck
CAS Number:  
Irritant
Sulpho-LC-SDA (Sulpho-NHS-LC-Diazirine) (Sulphosuccinimidyl 6-(4,4′-azipentanamido)hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803359-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :440.40 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Refrigerator
Formula:C₁₅H₂₁N₄NaO₈S
MDL Number:MFCD28899432