LC-SDA (NHS-LC-Diazirine) (succinimidyl 6-(4,4′-azipentanamido)hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803421-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:338.36 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
Formula:C₁₅H₂₂N₄O₅