Abacavir Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1000420.
- 1000441.
- 1000452.
- 1000437.
- Description:
- Abacavir Related Compound A
- Abacavir Related Compound B
- Abacavir Related Compound C
- Abacavir Related Compound D
- Synonym(s):
- [4-(2,6-diamino-9H-purin-9-yl)cyclopent-2-enyl]methanol
- [4-(2,5-diamino-6-chloropyrimidin-4-ylamino)cyclopent-2-enyl]methanol
- [(1S,4R)-4-(2-amino-6-chloro-9H-purin-9-yl)cyclopent-2-enyl]methanol hydrochloride
- N6-Cyclopropyl-9-{(1R,4S)-4-[(2,5-diamino-6-chloropyrimidin-4-yloxy)methyl]cyclopent-2-enyl}-9H-purine-2,6-diamine
- CAS No.:
- 906626-51-5
- 172015-79-1
- 1443421-69-9
- Formula:
- C₁₁H₁₄N₆O
- Storage conditions:
- Refigerator
- Refigerator
- Refigerator
- Refigerator
Specifications
Product Family Options
Product Information
- StandardsPkAvailabilityPrice
- 1000420.Abacavir Related Compound ASpecifications:
More
Specifications
Cat. No.1000420.DescriptionAbacavir Related Compound ASynonym(s)[4-(2,6-diamino-9H-purin-9-yl)cyclopent-2-enyl]methanolCAS No.906626-51-5FormulaC₁₁H₁₄N₆OStorage conditionsRefigerator - 1000441.Abacavir Related Compound BSpecifications:
More
Specifications
Cat. No.1000441.DescriptionAbacavir Related Compound BSynonym(s)[4-(2,5-diamino-6-chloropyrimidin-4-ylamino)cyclopent-2-enyl]methanolCAS No.FormulaStorage conditionsRefigerator - 1000452.Abacavir Related Compound CSpecifications:
More
Specifications
Cat. No.1000452.DescriptionAbacavir Related Compound CSynonym(s)[(1S,4R)-4-(2-amino-6-chloro-9H-purin-9-yl)cyclopent-2-enyl]methanol hydrochlorideCAS No.172015-79-1FormulaStorage conditionsRefigerator - 1000437.Abacavir Related Compound DSpecifications:
More
Specifications
Cat. No.1000437.DescriptionAbacavir Related Compound DSynonym(s)N6-Cyclopropyl-9-{(1R,4S)-4-[(2,5-diamino-6-chloropyrimidin-4-yloxy)methyl]cyclopent-2-enyl}-9H-purine-2,6-diamineCAS No.1443421-69-9FormulaStorage conditionsRefigerator



