Order Entry
Export
Orders LinkContactUsLinkComponent
Piroxicam Related Compounds, Reference Standard

Piroxicam Related Compounds, Reference Standard

Supplier:   USP
Piroxicam Related Compounds, Reference Standard
Supplier:  USP
Certificates
Piroxicam Related Compounds, Reference Standard

5 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1544519.
  • 1544520.
  • 1544541.
  • 1544574.
  • 1544600.
  • Description:
  • Piroxicam Related Compound A
  • Piroxicam Related Compound B
  • Piroxicam Related Compound D
  • Piroxicam Related Compound G
  • Piroxicam Related Compound J
  • Synonym(s):
  • Pyridin-2-amine
  • 4-Hydroxy-N-(pyridin-2-yl)-2H-benzothiazine-3-carboxamide 1,1-dioxide
  • Methyl 2-[1,1-dioxido-3-oxobenzoisothiazol-2(3H)-yl]acetate
  • Methyl 4-hydroxy-2H-benzothiazine-3-carboxylate 1,1-dioxide monohydrate
  • Methyl 4-hydroxy-2-methyl-2H-benzothiazine-3-carboxylate 1,1-dioxide
  • CAS No.:
  • 504-29-0
  • 65897-46-3
  • 6639-62-9
  • 35511-15-0
  • UN No.:
  • 2671

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1544519.
    Piroxicam Related Compound A
    Piroxicam Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 080,00 €
    1 080,00 €
    1 * 50 mg
       1 080,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1544519.
    Description
    Piroxicam Related Compound A
    Synonym(s)
    Pyridin-2-amine
    CAS No.
    504-29-0
    UN No.
    2671
  • 1544520.
    Piroxicam Related Compound B
    Piroxicam Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 150,00 €
    1 150,00 €
    1 * 50 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1544520.
    Description
    Piroxicam Related Compound B
    Synonym(s)
    4-Hydroxy-N-(pyridin-2-yl)-2H-benzothiazine-3-carboxamide 1,1-dioxide
    CAS No.
    65897-46-3
    UN No.
  • 1544541.
    Piroxicam Related Compound D
    Piroxicam Related Compound D
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 230,00 €
    1 230,00 €
    1 * 50 mg
       1 230,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1544541.
    Description
    Piroxicam Related Compound D
    Synonym(s)
    Methyl 2-[1,1-dioxido-3-oxobenzoisothiazol-2(3H)-yl]acetate
    CAS No.
    6639-62-9
    UN No.
  • 1544574.
    Piroxicam Related Compound G
    Piroxicam Related Compound G
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 250,00 €
    1 250,00 €
    1 * 50 mg
       1 250,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1544574.
    Description
    Piroxicam Related Compound G
    Synonym(s)
    Methyl 4-hydroxy-2H-benzothiazine-3-carboxylate 1,1-dioxide monohydrate
    CAS No.
    UN No.
  • 1544600.
    Piroxicam Related Compound J
    Piroxicam Related Compound J
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 190,00 €
    1 190,00 €
    1 * 50 mg
       1 190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1544600.
    Description
    Piroxicam Related Compound J
    Synonym(s)
    Methyl 4-hydroxy-2-methyl-2H-benzothiazine-3-carboxylate 1,1-dioxide
    CAS No.
    35511-15-0
    UN No.