Ondansetron Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1478618.
- 1478629.
- 1478593.
- Description:
- Ondansetron Related Compound C
- Ondansetron Related Compound D
- Ondansetron Related Compound A
- Synonym(s):
- 1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one
- 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
- 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one hydrochloride
- CAS No.:
- 27387-31-1
- 99614-64-9
- 119812-29-2
- Formula:
- C₁₃H₁₃NO
- C₁₄H₁₃NO
- C₁₆H₂₁ClN₂O
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1478618.Ondansetron Related Compound COndansetron Related Compound CUSP reference standardVial30 mgLog in for availability1 * 30 mg1 240,00 €Specifications:
More
Specifications
Cat. No.1478618.DescriptionOndansetron Related Compound CSynonym(s)1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-oneCAS No.27387-31-1FormulaC₁₃H₁₃NOUN No.2811 - 1478629.Ondansetron Related Compound DOndansetron Related Compound DUSP reference standardVial30 mgLog in for availability1 * 30 mg1 260,00 €Specifications:
More
Specifications
Cat. No.1478629.DescriptionOndansetron Related Compound DSynonym(s)1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-oneCAS No.99614-64-9FormulaC₁₄H₁₃NOUN No. - 1478593.Ondansetron Related Compound AOndansetron Related Compound AUSP reference standardVial50 mgLog in for availability1 * 50 mg1 240,00 €Specifications:
More
Specifications
Cat. No.1478593.DescriptionOndansetron Related Compound ASynonym(s)3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one hydrochlorideCAS No.119812-29-2FormulaC₁₆H₂₁ClN₂OUN No.



