Order Entry
Export
ContactUsLinkComponent
Latanoprost Related Compounds, Reference Standard
Latanoprost Related Compounds, Reference Standard
Supplier:  USP
Certificates
Latanoprost Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    145773-22-4
  • Synonym(s):
    Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoate
  • Formula:
    C₂₆H₄₀O₅
  • Description:
    Latanoprost Related Compound B

Specifications

Related Information