Hydrocortisone Hemisuccinate, Reference Standard
1319002.
: USP
:
- Pack type:Vial
- Pk:200 mg
- Standards:USP reference standard
- CAS No.:83784-20-7
- Description:Hydrocortisone Hemisuccinate
- Seq:02
- Formula:C₂₅H₃₆O₉
- Description:Hydrocortisone Hemisuccinate
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard