Order Entry
Export
Orders LinkContactUsLinkComponent
Ecamsule Related Compounds, Reference Standard

Ecamsule Related Compounds, Reference Standard

Supplier:   USP
Ecamsule Related Compounds, Reference Standard
Supplier:  USP
Certificates
Ecamsule Related Compounds, Reference Standard

6 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1231615.
  • 1231626.
  • 1231637.
  • 1231648.
  • 1231659.
  • 1231681.
  • Description:
  • Ecamsule Related Compound A
  • Ecamsule Related Compound B
  • Ecamsule Related Compound C
  • Ecamsule Related Compound D
  • Ecamsule Related Compound E
  • Ecamsule Related Compound G
  • Synonym(s):
  • 1,4-Phenylenedimethanol
  • 4-(Hydroxymethyl)benzoic acid
  • terephthalic acid
  • ((1SR,4RS)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid
  • sodium ((1SR,4SR,E)-3-(4-(hydroxymethyl)benzylidene)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate
  • 4-((E)-((1SR,4SR)-7,7-dimethyl-3-oxo-4-(sulfomethyl)bicyclo[2.2.1]heptan-2-ylidene)methyl)benzaldehyde, sodium salt
  • CAS No.:
  • 589-29-7
  • 3006-96-0
  • 100-21-0
  • 5872-08-2
  • UN No.:
  • 3261

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1231615.
    Ecamsule Related Compound A
    Ecamsule Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 180,00 €
    1 180,00 €
    1 * 50 mg
       1 180,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231615.
    Description
    Ecamsule Related Compound A
    Synonym(s)
    1,4-Phenylenedimethanol
    CAS No.
    589-29-7
    UN No.
  • 1231626.
    Ecamsule Related Compound B
    Ecamsule Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 180,00 €
    1 180,00 €
    1 * 50 mg
       1 180,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231626.
    Description
    Ecamsule Related Compound B
    Synonym(s)
    4-(Hydroxymethyl)benzoic acid
    CAS No.
    3006-96-0
    UN No.
  • 1231637.
    Ecamsule Related Compound C
    Ecamsule Related Compound C
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 120,00 €
    1 120,00 €
    1 * 50 mg
       1 120,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231637.
    Description
    Ecamsule Related Compound C
    Synonym(s)
    terephthalic acid
    CAS No.
    100-21-0
    UN No.
  • 1231648.
    Ecamsule Related Compound D
    Ecamsule Related Compound D
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 541,00 €
    541,00 €
    1 * 50 mg
       541,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231648.
    Description
    Ecamsule Related Compound D
    Synonym(s)
    ((1SR,4RS)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid
    CAS No.
    5872-08-2
    UN No.
    3261
  • 1231659.
    Ecamsule Related Compound E
    Ecamsule Related Compound E
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 270,00 €
    1 270,00 €
    1 * 25 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231659.
    Description
    Ecamsule Related Compound E
    Synonym(s)
    sodium ((1SR,4SR,E)-3-(4-(hydroxymethyl)benzylidene)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate
    CAS No.
    UN No.
  • 1231681.
    Ecamsule Related Compound G
    Ecamsule Related Compound G
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 509,00 €
    509,00 €
    1 * 25 mg
       509,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1231681.
    Description
    Ecamsule Related Compound G
    Synonym(s)
    4-((E)-((1SR,4SR)-7,7-dimethyl-3-oxo-4-(sulfomethyl)bicyclo[2.2.1]heptan-2-ylidene)methyl)benzaldehyde, sodium salt
    CAS No.
    UN No.