Order Entry
Export
ContactUsLinkComponent
Doxazosin Related Compounds, Reference Standard
Doxazosin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Doxazosin Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    27631-29-4
  • Synonym(s):
    2,4-dichloro-6,7-dimethoxyquinazoline
  • Formula:
    C₁₀H₈Cl₂N₂O₂
  • Description:
    Doxazosin Related Compound E
  • UN No.:
    2811

Specifications