Clopidogrel Amide, Pharmaceutical Analytical Impurities
1A01860.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:444728-13-6
- Description:Clopidogrel Amide
- Synonym(s):(S)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetamide
- Description:Clopidogrel Amide
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.