Order Entry
Export
ContactUsLinkComponent
 
Zonisamide, Reference Standards
 :  USP
 
Zonisamide, Reference Standards
 :  USP

 

  • CAS No.:
    73101-64-1
  • Synonym(s):
    1,2-benzisoxazole-3-methanesulfonic acid sodium salt
  • Description:
    Zonisamide Related Compound A