Order Entry
Export
ContactUsLinkComponent
Terbutaline Related Compounds, Reference Standard
Terbutaline Related Compounds, Reference Standard
Supplier:  USP
Certificates
Terbutaline Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    94109-61-2
  • Synonym(s):
    2-[Benzyl(tert-butyl)amino]-1-(3,5-dihydroxyphenyl)ethan-1-one
  • Description:
    Terbutaline Related Compound D
  • Formula:
    C₂₄H₄₀N₂O₁₀S

Specifications