Saquinavir Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1609829.
- 1609831.
- Description:
- Saquinavir Mesylate
- Saquinavir Related Compound A
- Synonym(s):
- N-tert-butyl-decahydro-2-[2(R)-hydroxy-4-phenyl-3(S)-[[N-(2-quinolylcarbonyl)-D-asparaginyl]amino]butyl]-(4aS,8aS)-isoquinoline-3(S)-carboxamide
- CAS No.:
- 149845-06-7
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1609829.Saquinavir MesylateSpecifications:
More
Specifications
Cat. No.1609829.DescriptionSaquinavir MesylateSynonym(s)CAS No.149845-06-7 - 1609831.Saquinavir Related Compound ASaquinavir Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 190,00 €Specifications:
More
Specifications
Cat. No.1609831.DescriptionSaquinavir Related Compound ASynonym(s)N-tert-butyl-decahydro-2-[2(R)-hydroxy-4-phenyl-3(S)-[[N-(2-quinolylcarbonyl)-D-asparaginyl]amino]butyl]-(4aS,8aS)-isoquinoline-3(S)-carboxamideCAS No.



