Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ropinirole Related Compounds, Reference Standard

 :   USP
Ropinirole Related Compounds, Reference Standard
 :  USP
 
Ropinirole Related Compounds, Reference Standard

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1605205.
  • 1605216.
  • 1605227.
  • Description:
  • Ropinirole Hydrochloride
  • Ropinirole Related Compound A
  • Ropinirole Related Compound B
  • Synonym(s):
  • 4-(2-hydroxyethyl)indolin-2-one
  • 4-[2-(Dipropylamino)ethyl]indoline-2,3-dione hydrochloride
  • CAS No.:
  • 91374-20-8
  • 139122-19-3
  • 221264-21-7

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Ropinirole Hydrochloride
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - 1 110,00 €
    1 110,00 €
    1 * 200 mg
       1 110,00 €

     
     
    Specifications
    Cat. No.
    1605205.
    Description
    Ropinirole Hydrochloride
    Synonym(s)
    CAS No.
    91374-20-8
  • Ropinirole Related Compound A
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 700,00 €
    700,00 €
    1 * 50 mg
       700,00 €

     
     
    Specifications
    Cat. No.
    1605216.
    Description
    Ropinirole Related Compound A
    Synonym(s)
    4-(2-hydroxyethyl)indolin-2-one
    CAS No.
    139122-19-3
  • Ropinirole Related Compound B
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 1 240,00 €
    1 240,00 €
    1 * 50 mg
       1 240,00 €

     
     
    Specifications
    Cat. No.
    1605227.
    Description
    Ropinirole Related Compound B
    Synonym(s)
    4-[2-(Dipropylamino)ethyl]indoline-2,3-dione hydrochloride
    CAS No.
    221264-21-7