Order Entry
Export
Orders LinkContactUsLinkComponent
Ezetimibe, Pharmaceutical Analytical Impurities

Ezetimibe, Pharmaceutical Analytical Impurities

R,R,S-Ezetimibe, Pharmaceutical Analytical Impurities, Vial, 25 mg
Catalog # 1A00140.
Supplier:   USP
Ezetimibe, Pharmaceutical Analytical Impurities
Catalog # 1A00140.
Supplier:  USP
CAS Number:  
Ezetimibe, Pharmaceutical Analytical Impurities

R,R,S-Ezetimibe, Pharmaceutical Analytical Impurities, Vial, 25 mg

CAS Number:  
Price
958,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • Description:
    R,R,S-Ezetimibe
  • Cat. No.:
    1A00140.
  • Description:
    R,R,S-Ezetimibe
  • Synonym(s):
    (3R,4S)-1-(4-fluorophenyl)-3-((R)-3-(4-fluorophenyl)-3-hydroxypropyl)-4-(4-hydroxyphenyl)azetidin-2-one
  • CAS No.:
    163380-16-3

Specifications