Order Entry
Export
Orders LinkContactUsLinkComponent
Palonosetron, Reference Standards

Palonosetron, Reference Standards

 :   USP
Palonosetron, Reference Standards
 :  USP
 
Palonosetron, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1492200.
  • 1492211.
  • Description:
  • Palonosetron Hydrochloride
  • Palonosetron Enantiomer
  • Synonym(s):
  • (3aR)-2-[(3R)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride
  • CAS No.:
  • 135729-62-3
  • 135729-75-8

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Palonosetron Hydrochloride
    USP reference standard
    Vial
    100 mg
       
       
    1 * 100 mg
    1 * 100 mg - 406,00 €
    406,00 €
    1 * 100 mg
       406,00 €

     
     
    Specifications
    Cat. No.
    1492200.
    Description
    Palonosetron Hydrochloride
    Synonym(s)
    CAS No.
    135729-62-3
  • Palonosetron Enantiomer
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 1 210,00 €
    1 210,00 €
    1 * 15 mg
       1 210,00 €

     
     
    Specifications
    Cat. No.
    1492211.
    Description
    Palonosetron Enantiomer
    Synonym(s)
    (3aR)-2-[(3R)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride
    CAS No.
    135729-75-8