Order Entry
Export
Orders LinkContactUsLinkComponent
Oxacillin Related Compounds, Reference Standard

Oxacillin Related Compounds, Reference Standard

Supplier:   USP
Oxacillin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Oxacillin Related Compounds, Reference Standard

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1481000.
  • 1481044.
  • Description:
  • Oxacillin Sodium
  • Oxacillin Related Compound C
  • Synonym(s):
  • 5-Methyl-3-phenylisoxazole-4-carboxylic acid
  • CAS No.:
  • 7240-38-2
  • 1136-45-4

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1481000.
    Oxacillin Sodium
    Oxacillin Sodium
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 406,00 €
    406,00 €
    1 * 200 mg
       406,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1481000.
    Description
    Oxacillin Sodium
    Synonym(s)
    CAS No.
    7240-38-2
  • 1481044.
    Oxacillin Related Compound C
    Oxacillin Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 150,00 €
    1 150,00 €
    1 * 15 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1481044.
    Description
    Oxacillin Related Compound C
    Synonym(s)
    5-Methyl-3-phenylisoxazole-4-carboxylic acid
    CAS No.
    1136-45-4