Order Entry
Export
Orders LinkContactUsLinkComponent
Glycopyrrolate, Reference Standards

Glycopyrrolate, Reference Standards

Supplier:   USP
Glycopyrrolate, Reference Standards
Supplier:  USP
Certificates

7 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1296009.
  • 1296010.
  • 1296029.
  • 1296031.
  • 1296042.
  • 1296086.
  • 1296100.
  • Description:
  • Glycopyrrolate
  • Glycopyrrolate Erythro Isomer
  • Glycopyrrolate Related Compound A
  • Glycopyrrolate Related Compound B
  • Glycopyrrolate Related Compound C
  • Glycopyrrolate Related Compound I
  • Glycopyrrolate Related Compound L
  • Synonym(s):
  • (RS)-3-[(RS)-2-cyclopentyl-2-hydroxy-2-phenylacetoxy]-1,1-dimethylpyrrolidinium bromide
  • 5-Nitrobenzene-1,3-dicarboxylic acid
  • 1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate Hydrochloride
  • 2-Cyclopentyl-2-hydroxy-2-phenylacetic acid
  • (RS)-3-[(SR)-2-(4-Chlorophenyl)-2-cyclopentyl-2-hydroxyacetoxy]-1,1-dimethylpyrrolidin-1-ium bromide
  • Methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate
  • CAS No.:
  • 51186-83-5
  • 618-88-2
  • 13118-10-0
  • 427-49-6
  • 19833-96-6

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1296009.
    Glycopyrrolate
    Glycopyrrolate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 406,00 €
    406,00 €
    1 * 200 mg
       406,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296009.
    Description
    Glycopyrrolate
    Synonym(s)
    CAS No.
    51186-83-5
  • 1296010.
    Glycopyrrolate Erythro Isomer
    Glycopyrrolate Erythro Isomer
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 430,00 €
    1 430,00 €
    1 * 25 mg
       1 430,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296010.
    Description
    Glycopyrrolate Erythro Isomer
    Synonym(s)
    (RS)-3-[(RS)-2-cyclopentyl-2-hydroxy-2-phenylacetoxy]-1,1-dimethylpyrrolidinium bromide
    CAS No.
  • 1296029.
    Glycopyrrolate Related Compound A
    Glycopyrrolate Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 240,00 €
    1 240,00 €
    1 * 50 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296029.
    Description
    Glycopyrrolate Related Compound A
    Synonym(s)
    5-Nitrobenzene-1,3-dicarboxylic acid
    CAS No.
    618-88-2
  • 1296031.
    Glycopyrrolate Related Compound B
    Glycopyrrolate Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 260,00 €
    1 260,00 €
    1 * 50 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296031.
    Description
    Glycopyrrolate Related Compound B
    Synonym(s)
    1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate Hydrochloride
    CAS No.
    13118-10-0
  • 1296042.
    Glycopyrrolate Related Compound C
    Glycopyrrolate Related Compound C
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 240,00 €
    1 240,00 €
    1 * 25 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296042.
    Description
    Glycopyrrolate Related Compound C
    Synonym(s)
    2-Cyclopentyl-2-hydroxy-2-phenylacetic acid
    CAS No.
    427-49-6
  • 1296086.
    Glycopyrrolate Related Compound I
    Glycopyrrolate Related Compound I
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 360,00 €
    1 360,00 €
    1 * 15 mg
       1 360,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296086.
    Description
    Glycopyrrolate Related Compound I
    Synonym(s)
    (RS)-3-[(SR)-2-(4-Chlorophenyl)-2-cyclopentyl-2-hydroxyacetoxy]-1,1-dimethylpyrrolidin-1-ium bromide
    CAS No.
  • 1296100.
    Glycopyrrolate Related Compound L
    Glycopyrrolate Related Compound L
    USP reference standard
    Ampule
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 240,00 €
    1 240,00 €
    1 * 50 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1296100.
    Description
    Glycopyrrolate Related Compound L
    Synonym(s)
    Methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate
    CAS No.
    19833-96-6