Order Entry
Export
ContactUsLinkComponent
Glacial Acetic Acid, Reference Standard
Glacial Acetic Acid, Reference Standard
Catalog # 1005706.
Supplier:  USP
CAS Number:  
undefined
Glacial Acetic Acid, Reference Standard
Catalog # 1005706.
Supplier:  USP
CAS Number:  

Specifications

  • Pk:
    3
  • Standards:
    USP reference standard
  • CAS No.:
    64-19-7
  • Description:
    Glacial Acetic Acid
  • UN No.:
    2789
  • Formula:
    C₂H₄O₂
  • Description:
    Glacial Acetic Acid
  • Pk Info:
    3 x 1.5 mL ampules

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard