Order Entry
Export
Orders LinkContactUsLinkComponent
Fenbendazole, Reference Standards

Fenbendazole, Reference Standards

 :   USP
Fenbendazole, Reference Standards
 :  USP
 
Fenbendazole, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1269403.
  • 1269414.
  • 1269425.
  • Description:
  • Fenbendazole
  • Fenbendazole Related Compound A
  • Fenbendazole Related Compound B
  • Synonym(s):
  • Methyl (1H-benzimidazole-2-yl)carbamate
  • Methyl [5(6)-chlorobenzimidazole-2-yl]carbamate
  • CAS No.:
  • 43210-67-9
  • 10605-21-7
  • 20367-38-8
  • UN No.:
  • 3077

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Fenbendazole
    USP reference standard
    Vial
    100 mg
       
       
    1 * 100 mg
    1 * 100 mg - 1 070,00 €
    1 070,00 €
    1 * 100 mg
       1 070,00 €

     
     
    Specifications
    Cat. No.
    1269403.
    Description
    Fenbendazole
    Synonym(s)
    CAS No.
    43210-67-9
    UN No.
  • Fenbendazole Related Compound A
    USP reference standard
    Vial
    30 mg
       
       
    1 * 30 mg
    1 * 30 mg - 1 210,00 €
    1 210,00 €
    1 * 30 mg
       1 210,00 €

     
     
    Specifications
    Cat. No.
    1269414.
    Description
    Fenbendazole Related Compound A
    Synonym(s)
    Methyl (1H-benzimidazole-2-yl)carbamate
    CAS No.
    10605-21-7
    UN No.
    3077
  • Fenbendazole Related Compound B
    USP reference standard
    Vial
    30 mg
       
       
    1 * 30 mg
    1 * 30 mg - 1 180,00 €
    1 180,00 €
    1 * 30 mg
       1 180,00 €

     
     
    Specifications
    Cat. No.
    1269425.
    Description
    Fenbendazole Related Compound B
    Synonym(s)
    Methyl [5(6)-chlorobenzimidazole-2-yl]carbamate
    CAS No.
    20367-38-8
    UN No.