Order Entry
Export
Orders LinkContactUsLinkComponent
Dipyridamole, Reference Standards

Dipyridamole, Reference Standards

Supplier:   USP
Dipyridamole, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1220506.
  • 1220561.
  • Description:
  • Dipyridamole
  • Dipyridamole
  • Synonym(s):
  • 2,2',2'',2'''-({4-[(2-Hydroxyethyl)amino]-8-(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl}bis(azanetriyl))tetraethanol
  • CAS No.:
  • 58-32-2
  • 60286-30-8

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • Dipyridamole
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 372,00 €
    372,00 €
    1 * 200 mg
       372,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1220506.
    Description
    Dipyridamole
    Synonym(s)
    CAS No.
    58-32-2
  • 1220561.
    Dipyridamole
    Dipyridamole
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 190,00 €
    1 190,00 €
    1 * 25 mg
       1 190,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1220561.
    Description
    Dipyridamole
    Synonym(s)
    2,2',2'',2'''-({4-[(2-Hydroxyethyl)amino]-8-(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl}bis(azanetriyl))tetraethanol
    CAS No.
    60286-30-8